siRNA Details
Gene Symbol: LOC101929450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125814.1 | 4 | 1715 | |||
NR_125815.1 | 4 | 2214 | ||||
GenBank mRNA | BC015784.1 | - | 4 | 2169 | ||
BC047326.1 | - | 4 | 2173 | |||
BC082237.1 | - | 4 | 1699 |
