siRNA Details
| Gene Symbol: C9orf139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207511.2 | NP_997394.1 | 3 | 3693 | ||
| XM_017014717.1 | XP_016870206.1 | 3 | 2658 | |||
| GenBank mRNA | AK093587.1 | - | 2 | 2062 | ||
| AK124899.1 | - | 3 | 3693 | |||

C9orf139
Genomic Map