siRNA Details
| Gene Symbol: LOC101929340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120577.1 | 2 | 1052 | |||
| NR_120579.1 | 2 | 817 | ||||
| GenBank mRNA | AK000493.1 | - | 2 | 1058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929340| Gene Symbol: LOC101929340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120577.1 | 2 | 1052 | |||
| NR_120579.1 | 2 | 817 | ||||
| GenBank mRNA | AK000493.1 | - | 2 | 1058 | ||