siRNA Details
| Gene Symbol: LOC101927274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110751.1 | 2 | 1025 | |||
| GenBank mRNA | BC039326.1 | - | 2 | 1029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927274| Gene Symbol: LOC101927274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110751.1 | 2 | 1025 | |||
| GenBank mRNA | BC039326.1 | - | 2 | 1029 | ||