siRNA Details
| Gene Symbol: CASC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125366.1 | 1 | 400 | |||
| GenBank mRNA | BC027619.2 | - | 1 | 380 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG771537.1 | - | 1 | 385 | Chr. 11 - 8012251 - 8016489 [-] on Build GRCh38 | |

CASC23
Genomic Map