siRNA Details
| Gene Symbol: NADK2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC033402.1 | - | 2 | 666 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NADK2-AS1| Gene Symbol: NADK2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC033402.1 | - | 2 | 666 | ||