siRNA Details
| Gene Symbol: LOC101927616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110057.1 | 1 | 72 | |||
| GenBank mRNA | BC039530.1 | - | 1 | 70 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM564278.1 | - | 1 | 90 | Chr. 12 - 127324155 - 127340072 [-] on Build GRCh38 | |

LOC101927616
Genomic Map