siRNA Details
| Gene Symbol: HNCAT21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104609.1 | 2 | 849 | |||
| GenBank mRNA | BC036254.1 | - | 2 | 849 | ||
| BC044609.1 | - | 2 | 848 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HNCAT21| Gene Symbol: HNCAT21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104609.1 | 2 | 849 | |||
| GenBank mRNA | BC036254.1 | - | 2 | 849 | ||
| BC044609.1 | - | 2 | 848 | |||