siRNA Details
| Gene Symbol: LOC100996664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135816.1 | 2 | 452 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI791647.1 | - | 2 | 110 | Chr. 14 - 68627181 - 68628445 [-] on Build GRCh38 | |

LOC100996664
Genomic Map