siRNA Details
| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 8 | 2287 | |||
| GenBank mRNA | AL137411.1 | - | 12 | 1281 | ||
| BC045761.1 | - | 8 | 2286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTE2P6| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 8 | 2287 | |||
| GenBank mRNA | AL137411.1 | - | 12 | 1281 | ||
| BC045761.1 | - | 8 | 2286 | |||