siRNA Details
| Gene Symbol: LINC01028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104129.1 | 3 | 834 | |||
| NR_104130.1 | 2 | 712 | ||||
| GenBank mRNA | BX647864.1 | - | 2 | 707 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG185376.1 | - | 1 | 577 | Chr. 17 - 70067181 - 70067964 [+] on Build GRCh38 | |

LINC01028
Genomic Map