siRNA Details
| Gene Symbol: LOC101928335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110395.1 | 4 | 1401 | |||
| GenBank mRNA | AK094711.1 | - | 4 | 1401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928335| Gene Symbol: LOC101928335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110395.1 | 4 | 1401 | |||
| GenBank mRNA | AK094711.1 | - | 4 | 1401 | ||