siRNA Details
Gene Symbol: LOC100506688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104614.1 | 2 | 2092 | |||
NR_104615.1 | 2 | 2026 | ||||
GenBank mRNA | AK126199.1 | - | 2 | 2060 | ||
AK127451.1 | - | 1 | 1226 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104614.1 | 2 | 2092 | |||
NR_104615.1 | 2 | 2026 | ||||
GenBank mRNA | AK126199.1 | - | 2 | 2060 | ||
AK127451.1 | - | 1 | 1226 |