siRNA Details
| Gene Symbol: TEX36-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023362.1 | 4, 5 | 508 | |||
| GenBank mRNA | BC033977.2 | - | 4, 5 | 508 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX36-AS1| Gene Symbol: TEX36-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023362.1 | 4, 5 | 508 | |||
| GenBank mRNA | BC033977.2 | - | 4, 5 | 508 | ||