siRNA Details
Gene Symbol: LOC100506444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125918.1 | 3 | 1264 | |||
GenBank mRNA | AK129929.1 | - | 3 | 1264 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125918.1 | 3 | 1264 | |||
GenBank mRNA | AK129929.1 | - | 3 | 1264 |