siRNA Details
| Gene Symbol: TTC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066351.1 | - | 2 | 414 | ||
| BF979681.2 | - | 419 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC3-AS1| Gene Symbol: TTC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066351.1 | - | 2 | 414 | ||
| BF979681.2 | - | 419 | ||||