siRNA Details
Gene Symbol: LOC101927787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125944.1 | 4 | 1345 | |||
GenBank mRNA | BC032040.1 | - | 4 | 1345 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125944.1 | 4 | 1345 | |||
GenBank mRNA | BC032040.1 | - | 4 | 1345 |