siRNA Details
| Gene Symbol: LINC01184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015360.1 | 2 | 192 | |||
| GenBank mRNA | AF088062.1 | - | 2 | 145 | ||
| AK090949.1 | - | 2 | 960 | |||
| BC036629.2 | - | 2 | 453 | |||
| BX648677.1 | - | 2 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01184| Gene Symbol: LINC01184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015360.1 | 2 | 192 | |||
| GenBank mRNA | AF088062.1 | - | 2 | 145 | ||
| AK090949.1 | - | 2 | 960 | |||
| BC036629.2 | - | 2 | 453 | |||
| BX648677.1 | - | 2 | 192 | |||