siRNA Details
| Gene Symbol: LINC00703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108054.1 | 3 | 605 | |||
| NR_108055.1 | 2 | 502 | ||||
| NR_108056.1 | 2 | 375 | ||||
| GenBank mRNA | AK095699.1 | - | 2 | 502 | ||
| BX095585.1 | - | 2 | 375 | |||
| DA500285.1 | - | 3 | 430 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA515194.1 | - | 2 | 502 | Chr. 10 - 4384246 - 4408772 [+] on Build GRCh38 | |

LINC00703
Genomic Map