siRNA Details
| Gene Symbol: LOC101927087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110838.1 | 9 | 763 | |||
| GenBank mRNA | AK057455.1 | - | 9 | 763 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927087| Gene Symbol: LOC101927087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110838.1 | 9 | 763 | |||
| GenBank mRNA | AK057455.1 | - | 9 | 763 | ||