siRNA Details
| Gene Symbol: LOC101929297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125862.1 | 2 | 471 | |||
| GenBank mRNA | BC040873.1 | - | 2 | 471 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM906286.1 | - | 2 | 490 | Chr. 6 - 166236967 - 166243625 [+] on Build GRCh38 | |

LOC101929297
Genomic Map