siRNA Details
| Gene Symbol: SMIM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104064.1 | 1, 2 | 367 | |||
| GenBank mRNA | DB036693.1 | - | 1, 2 | 367 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA399284.1 | - | 1, 2 | 58 | Chr. 13 - 44142637 - 44148028 [+] on Build GRCh38 | |

SMIM2-AS1
Genomic Map