siRNA Details
| Gene Symbol: LOC101927009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135195.1 | 3 | 3525 | |||
| GenBank mRNA | CR749525.1 | - | 3 | 3528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927009| Gene Symbol: LOC101927009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135195.1 | 3 | 3525 | |||
| GenBank mRNA | CR749525.1 | - | 3 | 3528 | ||