siRNA Details
| Gene Symbol: EPN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_048576.1 | 1 | 104 | |||
| GenBank mRNA | BC037786.2 | - | 1 | 104 | ||
| BG705989.1 | - | 1 | 109 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPN2-AS1| Gene Symbol: EPN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_048576.1 | 1 | 104 | |||
| GenBank mRNA | BC037786.2 | - | 1 | 104 | ||
| BG705989.1 | - | 1 | 109 | |||