siRNA Details
| Gene Symbol: LOC101927282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110753.1 | 5 | 553 | |||
| GenBank mRNA | BC035172.1 | - | 5 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927282| Gene Symbol: LOC101927282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110753.1 | 5 | 553 | |||
| GenBank mRNA | BC035172.1 | - | 5 | 560 | ||