siRNA Details
| Gene Symbol: LOC101927359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125885.1 | 9 | 974 | |||
| GenBank mRNA | BC040219.1 | - | 9 | 971 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927359| Gene Symbol: LOC101927359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125885.1 | 9 | 974 | |||
| GenBank mRNA | BC040219.1 | - | 9 | 971 | ||