siRNA Details
| Gene Symbol: LOC101929352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134478.1 | 3 | 910 | |||
| GenBank mRNA | AK125507.1 | - | 2 | 1869 | ||
| BG181462.1 | - | 2 | 591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929352| Gene Symbol: LOC101929352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134478.1 | 3 | 910 | |||
| GenBank mRNA | AK125507.1 | - | 2 | 1869 | ||
| BG181462.1 | - | 2 | 591 | |||