siRNA Details
| Gene Symbol: LOC101927637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120451.1 | 4 | 574 | |||
| GenBank mRNA | BU567943.1 | - | 3 | 524 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX103747.1 | - | 3 | 496 | Chr. 12 - 127631309 - 127636106 [-] on Build GRCh38 | |

LOC101927637
Genomic Map