siRNA Details
| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038356.1 | - | 536 | |||
| Gene Symbol: LOC101928567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110839.1 | 3 | 618 | |||

UBE2R2
Genomic Map