siRNA Details
Gene Symbol: LINC00303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097662.1 | - | 3 | 624 | ||
BC104010.2 | - | 3 | 623 | 40024418 | ||
BC104012.2 | - | 3 | 623 | 40024422 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097662.1 | - | 3 | 624 | ||
BC104010.2 | - | 3 | 623 | 40024418 | ||
BC104012.2 | - | 3 | 623 | 40024422 |