siRNA Details
| Gene Symbol: LOC101927159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110004.1 | 4 | 1387 | |||
| GenBank mRNA | BC040166.1 | - | 4 | 1387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927159| Gene Symbol: LOC101927159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110004.1 | 4 | 1387 | |||
| GenBank mRNA | BC040166.1 | - | 4 | 1387 | ||