siRNA Details
Gene Symbol: TDRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024015.1 | 2 | 889 | |||
GenBank mRNA | BC033995.2 | - | 2 | 976 | ||
BC042123.1 | - | 2 | 976 | |||
DQ168992.1 | - | 2 | 889 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL045589.1 | - | 2 | 683 | Chr. 6 - 40378663 - 40379717 [+] on Build GRCh38 |
