siRNA Details
| Gene Symbol: LOC151121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122040.1 | 5 | 2111 | |||
| NR_122041.1 | 4 | 2023 | ||||
| NR_122042.1 | 4 | 2034 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC151121| Gene Symbol: LOC151121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122040.1 | 5 | 2111 | |||
| NR_122041.1 | 4 | 2023 | ||||
| NR_122042.1 | 4 | 2034 | ||||