siRNA Details
| Gene Symbol: ZNF841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026662.1 | XP_016882151.1 | 8 | 4879 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF841| Gene Symbol: ZNF841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026662.1 | XP_016882151.1 | 8 | 4879 | ||