siRNA Details
| Gene Symbol: LOC100506289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537871.1 | - | 2 | 1568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506289| Gene Symbol: LOC100506289 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537871.1 | - | 2 | 1568 | ||