siRNA Details
| Gene Symbol: LOC105374177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135546.1 | 3 | 1384 | |||
| GenBank mRNA | BC038760.1 | - | 3 | 1382 | ||
| DB507682.1 | - | 3 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374177| Gene Symbol: LOC105374177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135546.1 | 3 | 1384 | |||
| GenBank mRNA | BC038760.1 | - | 3 | 1382 | ||
| DB507682.1 | - | 3 | 333 | |||