siRNA Details
| Gene Symbol: LINC01353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_117097.1 | 2 | 265 | |||
| NR_117098.1 | 2 | 237 | ||||
| GenBank mRNA | CK902257.1 | - | 2 | 237 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ777852.1 | - | 2 | 167 | Chr. 1 - 203287770 - 203288534 [+] on Build GRCh38 | |

LINC01353
Genomic Map