siRNA Details
| Gene Symbol: LINC00163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033840.1 | 2 | 1608 | |||
| GenBank mRNA | BC036902.1 | - | 2 | 1744 | ||
| BC046564.1 | - | 2 | 1713 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00163| Gene Symbol: LINC00163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033840.1 | 2 | 1608 | |||
| GenBank mRNA | BC036902.1 | - | 2 | 1744 | ||
| BC046564.1 | - | 2 | 1713 | |||