siRNA Details
Gene Symbol: LOC101929089 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120572.1 | 4 | 507 | |||
NR_120573.1 | 3 | 400 | ||||
NR_120575.1 | 3 | 420 | ||||
NR_120576.1 | 3 | 232 | ||||
GenBank mRNA | AI885123.1 | - | 3 | 175 | ||
BE671460.1 | - | 4 | 301 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI458517.1 | - | 3 | 175 | Chr. 11 - 118519479 - 118530550 [-] on Build GRCh38 |
