siRNA Details
Gene Symbol: LINC00354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046992.1 | 3 | 317 | |||
NR_120401.1 | 4 | 453 | ||||
GenBank mRNA | BX117816.1 | - | 3 | 309 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046992.1 | 3 | 317 | |||
NR_120401.1 | 4 | 453 | ||||
GenBank mRNA | BX117816.1 | - | 3 | 309 |