siRNA Details
Gene Symbol: LINC01551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026731.1 | 2, 3 | 233 | |||
GenBank mRNA | BX248251.1 | - | 2, 3 | 140 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026731.1 | 2, 3 | 233 | |||
GenBank mRNA | BX248251.1 | - | 2, 3 | 140 |