siRNA Details
| Gene Symbol: LINC01233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110664.1 | 1, 2 | 294 | |||
| GenBank mRNA | HY002653.1 | - | 1, 2 | 249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01233| Gene Symbol: LINC01233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110664.1 | 1, 2 | 294 | |||
| GenBank mRNA | HY002653.1 | - | 1, 2 | 249 | ||