siRNA Details
| Gene Symbol: LOC100996654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110040.1 | 3 | 416 | |||
| GenBank mRNA | HY038517.1 | - | 3 | 417 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX108678.1 | - | 3 | 304 | Chr. 7 - 54759425 - 54804928 [+] on Build GRCh38 | |

LOC100996654
Genomic Map