siRNA Details
Gene Symbol: LOC100996654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110040.1 | 1, 2 | 220 | |||
GenBank mRNA | HY038517.1 | - | 1, 2 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX108678.1 | - | 1, 2 | 108 | Chr. 7 - 54759425 - 54804928 [+] on Build GRCh38 |
