siRNA Details
| Gene Symbol: LOC105374060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135547.1 | 4 | 504 | |||
| GenBank mRNA | BX092438.1 | - | 4 | 384 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA446499.1 | - | 4 | 384 | Chr. 3 - 118508432 - 118559075 [-] on Build GRCh38 | |

LOC105374060
Genomic Map