siRNA Details
| Gene Symbol: LOC102723661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125991.1 | 5 | 614 | |||
| GenBank mRNA | BX095322.1 | - | 5 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723661| Gene Symbol: LOC102723661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125991.1 | 5 | 614 | |||
| GenBank mRNA | BX095322.1 | - | 5 | 614 | ||