siRNA Details
| Gene Symbol: LOC101928880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110944.1 | 2, 3 | 349 | |||
| GenBank mRNA | BX110403.1 | - | 2, 3 | 349 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928880| Gene Symbol: LOC101928880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110944.1 | 2, 3 | 349 | |||
| GenBank mRNA | BX110403.1 | - | 2, 3 | 349 | ||