siRNA Details
Gene Symbol: MIR4290HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038882.1 | 3 | 1774 | |||
GenBank mRNA | BC039331.1 | - | 3 | 1773 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIR4290HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038882.1 | 3 | 1774 | |||
GenBank mRNA | BC039331.1 | - | 3 | 1773 |