siRNA Details
| Gene Symbol: HRAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104613.1 | 2 | 193 | |||
| GenBank mRNA | BU930695.1 | - | 2 | 182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HRAT5| Gene Symbol: HRAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104613.1 | 2 | 193 | |||
| GenBank mRNA | BU930695.1 | - | 2 | 182 | ||