siRNA Details
Gene Symbol: LINC01559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036555.1 | 2 | 360 | |||
GenBank mRNA | AK091129.1 | - | 2 | 326 | ||
AK307179.1 | - | 2 | 232 | |||
BC101220.1 | - | 2 | 177 | 40022214 | ||
BC101222.2 | - | 2 | 177 | 40022216 | ||
BC101223.1 | - | 2 | 177 | 40022217 | ||
BG421494.1 | - | 361 |
